COMH4211B312 Combi-Blocks, Inc.

4-Bromo-1-iodo-2-isopropoxybenzene

  • MDL number MFCD12547879
  • Cas Number : 1208075-71-1
  • Molecular Weight : 341.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4648

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4648
Availability details for
Quantity
  • Availability
  • Add to Cart