COMH4211B313 Combi-Blocks, Inc.

5-Chloro-2-(4-methoxy-benzylsulfanyl)-phenylamine

  • MDL number MFCD11128876
  • Cas Number : 1019361-65-9
  • Molecular Weight : 279.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4652

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4652
Availability details for
Quantity
  • Availability
  • Add to Cart