COMH4211B314 Combi-Blocks, Inc.

Diethyl (4-bromo-2-chlorobenzyl)phosphonate

  • MDL number MFCD29904685
  • Cas Number : 183137-30-6
  • Molecular Weight : 341.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4653

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4653
Availability details for
Quantity
  • Availability
  • Add to Cart