COMH4211B316 Combi-Blocks, Inc.

Octanoic acid, 8-[[(1,1-dimethylethoxy)carbonyl]amino]-, 2,5-dioxo-1-pyrrolidinyl ester

  • MDL number MFCD31381693
  • Cas Number : 149414-80-2
  • Molecular Weight : 356.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4656

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4656
Availability details for
Quantity
  • Availability
  • Add to Cart