COMH4211B317 Combi-Blocks, Inc.

1-(2,2,2-Trifluoroethyl)piperidine-4-carboxylic acid

  • MDL number MFCD08696500
  • Cas Number : 937602-76-1
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4659

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4659
Availability details for
Quantity
  • Availability
  • Add to Cart