COMH4211B319 Combi-Blocks, Inc.

1-(1-Isobutyl-2,3-dihydro-1h-indol-5-yl)-ethanone

  • MDL number MFCD31557847
  • Cas Number : 1601044-30-7
  • Molecular Weight : 217.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4666

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4666
Availability details for
Quantity
  • Availability
  • Add to Cart