COMH4211B31A Combi-Blocks, Inc.

2-Fluoro-4-(4-methanesulfonyl-piperazin-1-yl)-benzamide hydrochloride

  • MDL number MFCD31583025
  • Cas Number : 2270912-59-7
  • Molecular Weight : 337.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4667

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4667
Availability details for
Quantity
  • Availability
  • Add to Cart