COMH4211B31B Combi-Blocks, Inc.

1-(3-Bromo-phenyl)-4-oxo-1,4-dihydro-[1,8]naphthyridine-3-carboxylic acid (2,2,2-trifluoro-ethyl)-amide

  • MDL number MFCD31579567
  • Cas Number : 799803-99-9
  • Molecular Weight : 426.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4668

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4668
Availability details for
Quantity
  • Availability
  • Add to Cart