COMH4211B31C Combi-Blocks, Inc.

4-(4-Bromo-2-fluorobenzoyl)piperazine

  • MDL number MFCD09943319
  • Cas Number : 1017041-43-8
  • Molecular Weight : 287.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4671

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4671
Availability details for
Quantity
  • Availability
  • Add to Cart