COMH4211B31D Combi-Blocks, Inc.

1-(3-Trifluoromethyl-benzyl)-1h-pyrazol-4-ol

  • MDL number MFCD27871720
  • Cas Number : 1715776-50-3
  • Molecular Weight : 242.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4672

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4672
Availability details for
Quantity
  • Availability
  • Add to Cart