COMH4211B31E Combi-Blocks, Inc.

Ethyl 2-(6-bromo-2-naphthyloxy)acetate

  • MDL number MFCD00245482
  • Cas Number : 153291-12-4
  • Molecular Weight : 309.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4673

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4673
Availability details for
Quantity
  • Availability
  • Add to Cart