COMH4211B31F Combi-Blocks, Inc.

[2-(2-Prop-2-ynyloxy-ethoxy)-ethyl]-phosphonic acid diethyl ester

  • MDL number MFCD31613943
  • Cas Number : 1714139-58-8
  • Molecular Weight : 264.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name diethyl (2-(2-(prop-2-yn-1-yloxy)ethoxy)ethyl)phosphonate
Inchi Code 1S/C11H21O5P/c1-4-7-13-8-9-14-10-11-17(12,15-5-2)16-6-3/h1H,5-11H2,2-3H3
Inchi Key NGFJEWPAZHSGHR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4674

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4674
Availability details for
Quantity
  • Availability
  • Add to Cart