COMH4211B321 Combi-Blocks, Inc.

2-Chloro-2'-fluoro-3,4'-bipyridine

  • MDL number MFCD13191924
  • Cas Number : 870221-45-7
  • Molecular Weight : 208.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4677

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4677
Availability details for
Quantity
  • Availability
  • Add to Cart