COMH4211B322 Combi-Blocks, Inc.

tert-Butyl (3-amino-4-(tert-butyl)phenyl)carbamate

  • MDL number MFCD13182471
  • Cas Number : 873055-92-6
  • Molecular Weight : 264.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4680

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4680
Availability details for
Quantity
  • Availability
  • Add to Cart