COMH4211B323 Combi-Blocks, Inc.

2-[1-(Propan-2-yl)-1h-pyrazol-4-yl]oxyacetonitrile

  • MDL number MFCD29499194
  • Cas Number : 1597166-80-7
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4686

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4686
Availability details for
Quantity
  • Availability
  • Add to Cart