COMH4211B325 Combi-Blocks, Inc.

3-Bromo-5-((3,3-difluoropyrrolidin-1-yl)methyl)pyridine

  • MDL number MFCD30529994
  • Cas Number : 1428065-38-6
  • Molecular Weight : 277.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4692

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4692
Availability details for
Quantity
  • Availability
  • Add to Cart