COMH4211B326 Combi-Blocks, Inc.

tert-Butyl trans-4-(3-hydroxypropyl)-cyclohexylcarbamate

  • MDL number MFCD03844623
  • Cas Number : 1212142-46-5
  • Molecular Weight : 257.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4693

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4693
Availability details for
Quantity
  • Availability
  • Add to Cart