COMH4211B327 Combi-Blocks, Inc.

1-(4-Bromophenylsulfonyl)-4,4-difluoropiperidine

  • MDL number MFCD31583300
  • Cas Number : 1628016-71-6
  • Molecular Weight : 340.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4694

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4694
Availability details for
Quantity
  • Availability
  • Add to Cart