COMH4211B328 Combi-Blocks, Inc.

(5-Bromo-thiophen-2-ylmethyl)-[2-(3-fluoro-phenyl)-ethyl]-amine

  • MDL number MFCD12427977
  • Cas Number : 1007579-04-5
  • Molecular Weight : 314.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4699

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4699
Availability details for
Quantity
  • Availability
  • Add to Cart