COMH4211B329 Combi-Blocks, Inc.

Benzyl 5-formyl-1h-pyrrole-2-carboxylate

  • MDL number MFCD00598552
  • Cas Number : 183172-57-8
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4700

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4700
Availability details for
Quantity
  • Availability
  • Add to Cart