COMH4211B32B Combi-Blocks, Inc.

3-(4-Methylphenyl)butanoic acid

  • MDL number MFCD00231078
  • Cas Number : 39027-57-1
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4708

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4708
Availability details for
Quantity
  • Availability
  • Add to Cart