COMH4211B32C Combi-Blocks, Inc.

N-(4-Aminophenyl)-2-bromo-4-fluorobenzamide hydrochloride

  • MDL number MFCD31579517
  • Cas Number : 1578088-24-0
  • Molecular Weight : 345.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4710

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4710
Availability details for
Quantity
  • Availability
  • Add to Cart