COMH4211B330 Combi-Blocks, Inc.

5-Acetyl-2-benzyloxy-benzoic acid

  • MDL number MFCD22571420
  • Cas Number : 201663-18-5
  • Molecular Weight : 270.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4718

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4718
Availability details for
Quantity
  • Availability
  • Add to Cart