COMH4211B332 Combi-Blocks, Inc.

4-(4-Carboxy-3-fluoro-phenyl)-3,6-dihydro-2h-pyridine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31583072
  • Cas Number : 1643153-49-4
  • Molecular Weight : 321.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4725

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4725
Availability details for
Quantity
  • Availability
  • Add to Cart