COMH4211B333 Combi-Blocks, Inc.

[4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenoxy]-acetic acid tert-butyl ester

  • MDL number MFCD22494138
  • Cas Number : 769968-17-4
  • Molecular Weight : 334.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4726

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4726
Availability details for
Quantity
  • Availability
  • Add to Cart