COMH4211B334 Combi-Blocks, Inc.

2,4-Difluoro-5-iodo-benzoic acid tert-butyl ester

  • MDL number MFCD28975977
  • Cas Number : 1445862-86-1
  • Molecular Weight : 340.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4728

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4728
Availability details for
Quantity
  • Availability
  • Add to Cart