COMH4211B335 Combi-Blocks, Inc.

Ethyl 1-(6-aminopyridazin-3-yl)piperidine-4-carboxylate

  • MDL number MFCD31579513
  • Cas Number : 1770349-59-1
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4733

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4733
Availability details for
Quantity
  • Availability
  • Add to Cart