COMH4211B351 Combi-Blocks, Inc.

1-((4-Iodophenyl)sulfonyl)pyrrolidine

  • MDL number MFCD01993190
  • Cas Number : 167156-16-3
  • Molecular Weight : 337.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4745

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4745
Availability details for
Quantity
  • Availability
  • Add to Cart