COMH4211B354 Combi-Blocks, Inc.

Acetaldehyde, 2-[2-(benzoyloxy)ethoxy]-

  • MDL number MFCD25977887
  • Cas Number : 100420-34-6
  • Molecular Weight : 208.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4750

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4750
Availability details for
Quantity
  • Availability
  • Add to Cart