COMH4211B355 Combi-Blocks, Inc.

(4-Amino-bicyclo[2.2.2]oct-1-yl)-pyrrolidin-1-yl-methanone, hydrochloride

  • MDL number MFCD31539556
  • Cas Number : 2203714-64-9
  • Molecular Weight : 258.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4751

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4751
Availability details for
Quantity
  • Availability
  • Add to Cart