COMH4211B356 Combi-Blocks, Inc.

(2-Bromo-5-nitro-benzyl)-diethyl-amine

  • MDL number MFCD31437595
  • Cas Number : 1001057-06-2
  • Molecular Weight : 287.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4752

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4752
Availability details for
Quantity
  • Availability
  • Add to Cart