COMH4211B357 Combi-Blocks, Inc.

2-Bromo-1-(2,4-difluorophenoxy)-4-nitrobenzene

  • MDL number MFCD28406126
  • Cas Number : 183110-56-7
  • Molecular Weight : 330.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4753
Availability details for
Quantity
  • Availability
  • Add to Cart