COMH4211B358 Combi-Blocks, Inc.

N-Ethyl-2-methoxy-6-nitroaniline

  • MDL number MFCD31579424
  • Cas Number : 1416554-51-2
  • Molecular Weight : 196.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-ethyl-2-methoxy-6-nitroaniline
Inchi Code 1S/C9H12N2O3/c1-3-10-9-7(11(12)13)5-4-6-8(9)14-2/h4-6,10H,3H2,1-2H3
Inchi Key DTJVABSVCDSASD-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4754

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4754
Availability details for
Quantity
  • Availability
  • Add to Cart