COMH4211B359 Combi-Blocks, Inc.

(1-(((tert-Butyldiphenylsilyl)oxy)methyl)cyclopropyl)methanol

  • MDL number MFCD31619023
  • Cas Number : 441785-04-2
  • Molecular Weight : 340.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4755

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4755
Availability details for
Quantity
  • Availability
  • Add to Cart