COMH4211B35A Combi-Blocks, Inc.

4'-Bromo-3'-methyl-biphenyl-4-carboxylic acid methyl ester

  • MDL number MFCD31579460
  • Cas Number : 1411993-02-6
  • Molecular Weight : 305.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4756

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4756
Availability details for
Quantity
  • Availability
  • Add to Cart