COMH4211B799 Combi-Blocks, Inc.

5-Bromo-2-(2,2,2-trifluoro-ethoxy)-benzamide

  • MDL number MFCD31628795
  • Cas Number : 2270908-50-2
  • Molecular Weight : 298.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4758

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4758
Availability details for
Quantity
  • Availability
  • Add to Cart