COMH4211B79C Combi-Blocks, Inc.

4-Butoxy-2-chlorobenzoic acid

  • MDL number MFCD00599178
  • Cas Number : 95736-31-5
  • Molecular Weight : 228.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4768

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4768
Availability details for
Quantity
  • Availability
  • Add to Cart