COMH4211B79D Combi-Blocks, Inc.

3-[4-(4-Carbamoyl-3-fluoro-phenyl)-piperazin-1-yl]-propionic acid methyl ester

  • MDL number MFCD31583108
  • Cas Number : 2270912-71-3
  • Molecular Weight : 309.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4769

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4769
Availability details for
Quantity
  • Availability
  • Add to Cart