COMH4211B79E Combi-Blocks, Inc.

Ethyl 2-(dicyanomethyl)-2-methylbutanoate

  • MDL number MFCD31579445
  • Cas Number : 1838150-36-9
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4771

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4771
Availability details for
Quantity
  • Availability
  • Add to Cart