COMH4211B79F Combi-Blocks, Inc.

5-Bromo-n,n-dimethyl-2-(2,2,2-trifluoro-ethoxy)-benzamide

  • MDL number MFCD31628794
  • Cas Number : 2270910-39-7
  • Molecular Weight : 326.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4772

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4772
Availability details for
Quantity
  • Availability
  • Add to Cart