COMH4211B7A0 Combi-Blocks, Inc.

1-Bromo-2-fluoro-4-(2-fluoroethoxy)benzene

  • MDL number MFCD23970907
  • Cas Number : 1502762-25-5
  • Molecular Weight : 237.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4779

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4779
Availability details for
Quantity
  • Availability
  • Add to Cart