COMH4211B7A2 Combi-Blocks, Inc.

5-Bromo-n-cyclobutyl-3-methylpyridin-2-amine

  • MDL number MFCD23754307
  • Cas Number : 1289040-78-3
  • Molecular Weight : 241.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4781

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4781
Availability details for
Quantity
  • Availability
  • Add to Cart