COMH4211B7A4 Combi-Blocks, Inc.

1-(3-Fluoro-4-propoxyphenyl)ethanone

  • MDL number MFCD01861237
  • Cas Number : 203067-00-9
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4784

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4784
Availability details for
Quantity
  • Availability
  • Add to Cart