COMH4211B7A5 Combi-Blocks, Inc.

2-Amino-3-bromo-5-(4-trifluoromethylphenyl)pyridine

  • MDL number MFCD22379190
  • Cas Number : 911113-30-9
  • Molecular Weight : 317.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-5-[4-(trifluoromethyl)phenyl]-2-pyridinamine
Inchi Code 1S/C12H8BrF3N2/c13-10-5-8(6-18-11(10)17)7-1-3-9(4-2-7)12(14,15)16/h1-6H,(H2,17,18)
Inchi Key SVPYSCQEUIGBCD-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4785

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4785
Availability details for
Quantity
  • Availability
  • Add to Cart