COMH4211B7A6 Combi-Blocks, Inc.

tert-Butyl 4-(2-(hydroxymethyl)-4-nitrophenyl)piperazine-1-carboxylate

  • MDL number MFCD28964824
  • Cas Number : 955369-05-8
  • Molecular Weight : 337.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4786

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4786
Availability details for
Quantity
  • Availability
  • Add to Cart