COMH4211B7A8 Combi-Blocks, Inc.

N,2-Dimethyl-5-nitrobenzamide

  • MDL number MFCD19597437
  • Cas Number : 862470-06-2
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4788

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4788
Availability details for
Quantity
  • Availability
  • Add to Cart