COMH4211B7A9 Combi-Blocks, Inc.

6-Cyclopentyloxy-1h-indazol-3-ylamine

  • MDL number MFCD23111464
  • Cas Number : 1394934-96-3
  • Molecular Weight : 217.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4792

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4792
Availability details for
Quantity
  • Availability
  • Add to Cart