COMH4211B7AC Combi-Blocks, Inc.

4-Methyl-3-(1-methyl-1h-pyrazol-4-yl)aniline

  • MDL number MFCD18334191
  • Cas Number : 1342072-41-6
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4799

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4799
Availability details for
Quantity
  • Availability
  • Add to Cart