COMH4211B7AE Combi-Blocks, Inc.

5-Bromo-n-cyclopropyl-2-cyclopropylmethoxy-benzamide

  • MDL number MFCD31628801
  • Cas Number : 2270907-97-4
  • Molecular Weight : 310.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4801

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4801
Availability details for
Quantity
  • Availability
  • Add to Cart