COMH4211B7B0 Combi-Blocks, Inc.

3-(2-Fluorophenyl)-2-methylpropan-1-ol

  • MDL number MFCD20386023
  • Cas Number : 1342822-91-6
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4803

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4803
Availability details for
Quantity
  • Availability
  • Add to Cart